MAT062354371 Matrix Scientific

2,6-Naphthalenedithiol

  • MDL number MFCD00465086
  • Cas Number : 96892-95-4
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016608.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016608.html
Availability details for
Quantity
  • Availability
  • Add to Cart