MAT062354372 Matrix Scientific

2,7-Naphthalenedithiol

  • MDL number MFCD00068910
  • Cas Number : 71977-56-5
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016606.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016606.html
Availability details for
Quantity
  • Availability
  • Add to Cart