MAT062354418 Matrix Scientific

Phenethyl-phenyl-amine hydrochloride

  • MDL number MFCD03373261
  • Cas Number : 3441-11-0
  • Molecular Weight : 233.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Country of Origin USA
Product Link http://www.matrixscientific.com/008222.html

Safety Information

MSDS http://www.matrixscientific.com/008222.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart