MAT062354438 Matrix Scientific

1-(2-Phenylethyl)piperidin-3-amine sulfate

  • MDL number MFCD06801191
  • Cas Number : 1185299-91-5
  • Molecular Weight : 302.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016428.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016428.html
Availability details for
Quantity
  • Availability
  • Add to Cart