MAT062354439 Matrix Scientific

(2-Phenyl-1H-imidazol-1-yl)acetic acidhydrochloride

  • MDL number MFCD06801193
  • Cas Number : 1059626-11-7
  • Molecular Weight : 238.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016151.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016151.html
Availability details for
Quantity
  • Availability
  • Add to Cart