MAT062354452 Matrix Scientific

[(3-Phenyl-1H-pyrazol-4-yl)methyl]aminedihydrochloride

  • MDL number MFCD06801208
  • Cas Number : 936940-58-8
  • Molecular Weight : 246.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016546.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016546.html
Availability details for
Quantity
  • Availability
  • Add to Cart