MAT062354455 Matrix Scientific

4-Piperazinoaniline dihydrochloride

  • MDL number MFCD03131297
  • Cas Number : 1185303-28-9
  • Molecular Weight : 250.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007998.html

Safety Information

MSDS http://www.matrixscientific.com/007998.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart