MAT062354484 Matrix Scientific

4-(1H-Pyrazol-1-yl)benzoic acid hydrochloride

  • MDL number MFCD06801240
  • Cas Number : 1185294-89-6
  • Molecular Weight : 224.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016264.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016264.html
Availability details for
Quantity
  • Availability
  • Add to Cart