MAT062354492 Matrix Scientific

2-(2-Pyridin-3-ylethyl)benzoic acidhydrochloride

  • MDL number MFCD06801247
  • Cas Number : 1185303-88-1
  • Molecular Weight : 263.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016511.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016511.html
Availability details for
Quantity
  • Availability
  • Add to Cart