MAT062354497 Matrix Scientific

[4-(2-Pyridin-2-ylethyl)phenyl]aminedihydrochloride

  • MDL number MFCD06801252
  • Cas Number : 6318-71-4
  • Molecular Weight : 271.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57
Country of Origin USA
Product Link http://www.matrixscientific.com/016535.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016535.html
Availability details for
Quantity
  • Availability
  • Add to Cart