MAT062354514 Matrix Scientific

1-(Pyrimidin-2-yl)piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD06801270
  • Cas Number : 1185296-02-9
  • Molecular Weight : 243.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016497.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016497.html
Availability details for
Quantity
  • Availability
  • Add to Cart