MAT062354557 Matrix Scientific

2,3,7,8-Tetramethoxydibenzofuran

  • MDL number MFCD06656518
  • Cas Number : 76303-42-9
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016570.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016570.html
Availability details for
Quantity
  • Availability
  • Add to Cart