MAT062354572 Matrix Scientific

3,4,5-Trimethoxyphenylglyoxal hydrate

  • MDL number MFCD06801328
  • Cas Number : 858255-78-4
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/016082.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016082.html
Availability details for
Quantity
  • Availability
  • Add to Cart