MAT062354606 Matrix Scientific

4-(2-Methyl-thiazol-4-yl)-benzoic acidhydrobromide

  • MDL number MFCD01204078
  • Cas Number : 159887-01-1
  • Molecular Weight : 300.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010178.html

Safety Information

MSDS http://www.matrixscientific.com/010178.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart