MAT062354634 Matrix Scientific

1-(Piperidin-4-ylmethyl)piperidine

  • MDL number MFCD05215178
  • Cas Number : 32470-52-3
  • Molecular Weight : 182.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016343.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016343.html
Availability details for
Quantity
  • Availability
  • Add to Cart