MAT062354654 Matrix Scientific

2,7-Naphthalenedisulfonyl chloride

  • MDL number MFCD00277645
  • Cas Number : 19551-16-7
  • Molecular Weight : 325.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016605.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016605.html
Availability details for
Quantity
  • Availability
  • Add to Cart