MAT062354657 Matrix Scientific

4-n-Propylpiperidine

  • MDL number MFCD00023151
  • Cas Number : 22398-09-0
  • Molecular Weight : 127.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/016551.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/016551.html
Availability details for
Quantity
  • Availability
  • Add to Cart