MAT062354667 Matrix Scientific

N-Methyl-1-(1-methylpiperidin-3-yl)methanamine

  • MDL number MFCD05864531
  • Cas Number : 639078-61-8
  • Molecular Weight : 142.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016364.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016364.html
Availability details for
Quantity
  • Availability
  • Add to Cart