MAT062354669 Matrix Scientific

1-(4-Methyl-benzyl)-piperazinehydrochloride

  • MDL number MFCD05656272
  • Cas Number : 435345-17-8
  • Molecular Weight : 226.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008110.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008110.html
Availability details for
Quantity
  • Availability
  • Add to Cart