MAT062354726 Matrix Scientific

4-(Pyrrolidin-1-ylmethyl)piperidine

  • MDL number MFCD05215177
  • Cas Number : 683772-11-4
  • Molecular Weight : 168.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016344.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016344.html
Availability details for
Quantity
  • Availability
  • Add to Cart