MAT062354727 Matrix Scientific

(Octahydro-quinolizin-1-yl)-methanol

  • MDL number MFCD00017327
  • Cas Number : 10159-79-2
  • Molecular Weight : 169.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/016809.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016809.html
Availability details for
Quantity
  • Availability
  • Add to Cart