MAT062354729 Matrix Scientific

4-Tetrahydro-1H-pyrrol-3-ylpyridine

  • MDL number MFCD01862541
  • Cas Number : 150281-47-3
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 112�Torr
Product Link http://www.matrixscientific.com/016517.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016517.html
Availability details for
Quantity
  • Availability
  • Add to Cart