MAT062354776 Matrix Scientific

1,2,3,4-Tetramethoxybenzene

  • MDL number MFCD00068614
  • Cas Number : 21450-56-6
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/016632.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016632.html
Availability details for
Quantity
  • Availability
  • Add to Cart