MAT062354793 Matrix Scientific

4-(2-Phenoxyethoxy)aniline

  • MDL number MFCD00025327
  • Cas Number : 35965-96-9
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/017125.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/017125.html
Availability details for
Quantity
  • Availability
  • Add to Cart