MAT062354812 Matrix Scientific

2-(3,4,5-Trimethoxy-phenyl)thiazolidinehydrochloride

  • MDL number MFCD06801329
  • Cas Number : 1185296-81-4
  • Molecular Weight : 291.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007839.html

Safety Information

MSDS http://www.matrixscientific.com/007839.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart