MAT068370946 Matrix Scientific

4-Fluoro-3-methoxyphenylacetic acid

  • MDL number MFCD08146650
  • Cas Number : 946713-86-6
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/018258.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018258.html
Availability details for
Quantity
  • Availability
  • Add to Cart