MAT089755548 Matrix Scientific

4-p-Tolyl-piperidin-4-ol

  • MDL number MFCD00087127
  • Cas Number : 57988-60-0
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/018364.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018364.html
Availability details for
Quantity
  • Availability
  • Add to Cart