MAT089755549 Matrix Scientific

4-(4-Trifluoromethyl-phenyl)-piperidin-4-ol

  • MDL number MFCD08146665
  • Cas Number : 39757-71-6
  • Molecular Weight : 245.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018365.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018365.html
Availability details for
Quantity
  • Availability
  • Add to Cart