MAT089755587 Matrix Scientific

(Benzo[1,2,5]thiadiazol-4-yloxy)-acetic acid

  • MDL number MFCD00785106
  • Cas Number : 29289-27-8
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Country of Origin USA
Product Link http://www.matrixscientific.com/018403.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018403.html
Availability details for
Quantity
  • Availability
  • Add to Cart