MAT089755589 Matrix Scientific

4-(Pyrrolidine-1-sulfonyl)-benzoic acid

  • MDL number MFCD00551820
  • Cas Number : 19580-33-7
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018406.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018406.html
Availability details for
Quantity
  • Availability
  • Add to Cart