MAT089755591 Matrix Scientific

5-(2,2,6,6-Tetramethyl-1,2,3,6-tetrahydro-pyridin-4-yl)-thiophene-2-carbaldehyde

  • MDL number MFCD00507030
  • Cas Number : 99539-08-9
  • Molecular Weight : 249.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-92
Country of Origin USA
Product Link http://www.matrixscientific.com/018408.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018408.html
Availability details for
Quantity
  • Availability
  • Add to Cart