MAT089755603 Matrix Scientific

4-Oxo-2-pyrrolidin-1-yl-4H-pyrido[1,2-a]-pyrimidine-3-carbaldehyde

  • MDL number MFCD01072150
  • Cas Number : 111680-73-0
  • Molecular Weight : 243.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-145
Country of Origin USA
Product Link http://www.matrixscientific.com/018420.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018420.html
Availability details for
Quantity
  • Availability
  • Add to Cart