MAT089755605 Matrix Scientific

2-Amino-6-hydroxy-pyrimidine-4-carboxylic acid

  • MDL number MFCD00023261
  • Cas Number : 6973-52-0
  • Molecular Weight : 155.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018422.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018422.html
Availability details for
Quantity
  • Availability
  • Add to Cart