MAT089755610 Matrix Scientific

2-Benzylamino-N-biphenyl-2-yl-acetamide

  • MDL number MFCD01156955
  • Cas Number : 333341-34-7
  • Molecular Weight : 316.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018427.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018427.html
Availability details for
Quantity
  • Availability
  • Add to Cart