MAT089755619 Matrix Scientific

1-Allyl-1-pyridin-4-yl-but-3-enylamine

  • MDL number MFCD01455358
  • Cas Number : 304668-65-3
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018436.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018436.html
Availability details for
Quantity
  • Availability
  • Add to Cart