MAT089755635 Matrix Scientific

2-(1H-Indol-3-ylsulfanyl)-butyric acid

  • MDL number MFCD00981480
  • Cas Number : 164659-62-5
  • Molecular Weight : 235.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-80
Country of Origin USA
Product Link http://www.matrixscientific.com/018452.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018452.html
Availability details for
Quantity
  • Availability
  • Add to Cart