MAT089755644 Matrix Scientific

2-[4-(4-Methoxy-phenyl)-2,2-dimethyl-tetrahydro-pyran-4-yl]-ethylamine

  • MDL number MFCD01002967
  • Cas Number : 400748-66-5
  • Molecular Weight : 263.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018461.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018461.html
Availability details for
Quantity
  • Availability
  • Add to Cart