MAT089755683 Matrix Scientific

6-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]-indole

  • MDL number MFCD02127602
  • Cas Number : 350680-06-7
  • Molecular Weight : 186.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018500.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018500.html
Availability details for
Quantity
  • Availability
  • Add to Cart