MAT089755685 Matrix Scientific

(5-Amino-[1,3,4]thiadiazol-2-yl)-acetic acid

  • MDL number MFCD01346801
  • Cas Number : 118863-94-8
  • Molecular Weight : 159.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018502.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018502.html
Availability details for
Quantity
  • Availability
  • Add to Cart