MAT089755690 Matrix Scientific

1-(4-Benzyl-piperazin-1-yl)-2-chloro-ethanone

  • MDL number MFCD01964635
  • Cas Number : 56056-37-2
  • Molecular Weight : 252.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018508.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018508.html
Availability details for
Quantity
  • Availability
  • Add to Cart