MAT089755712 Matrix Scientific

3-Bromo-N-(4-pyridin-4-ylmethyl-phenyl)-propionamide

  • MDL number MFCD07186310
  • Cas Number : 842956-84-7
  • Molecular Weight : 319.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018530.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018530.html
Availability details for
Quantity
  • Availability
  • Add to Cart