MAT089755716 Matrix Scientific

2-Chloro-N-(5-methyl-benzo[1,2,5]thiadiazol-4-yl)-acetamide

  • MDL number MFCD02767285
  • Cas Number : 842957-18-0
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018534.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018534.html
Availability details for
Quantity
  • Availability
  • Add to Cart