MAT089755767 Matrix Scientific

2-[1-(2,3-Dihydro-benzo[1,4]dioxin-6-yl)-ethylamino]-ethanol

  • MDL number MFCD02999009
  • Cas Number : 383371-96-8
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018585.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018585.html
Availability details for
Quantity
  • Availability
  • Add to Cart