MAT089755768 Matrix Scientific

2-[1-(4-Ethoxy-phenyl)-ethylamino]-ethanol

  • MDL number MFCD04093197
  • Cas Number : 636997-13-2
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018586.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018586.html
Availability details for
Quantity
  • Availability
  • Add to Cart