MAT089755788 Matrix Scientific

4-(p-Tolylamino-methyl)-1H-quinolin-2-one

  • MDL number MFCD02575281
  • Cas Number : 333984-13-7
  • Molecular Weight : 264.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018606.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018606.html
Availability details for
Quantity
  • Availability
  • Add to Cart