MAT089755798 Matrix Scientific

1-(2-Diethylamino-ethyl)-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD06751683
  • Cas Number : 876708-73-5
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018616.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018616.html
Availability details for
Quantity
  • Availability
  • Add to Cart