MAT089755802 Matrix Scientific

3-(4-Oxo-4,5-dihydro-thiazol-2-ylamino)-benzoic acid

  • MDL number MFCD03043773
  • Cas Number : 432013-04-2
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018620.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018620.html
Availability details for
Quantity
  • Availability
  • Add to Cart