MAT089755826 Matrix Scientific

1-(4-Dimethylamino-phenyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD02604636
  • Cas Number : 169036-55-9
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018644.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018644.html
Availability details for
Quantity
  • Availability
  • Add to Cart