MAT089755843 Matrix Scientific

1-[1-(3,4-Dichloro-phenyl)-ethyl]-piperazine

  • MDL number MFCD05186464
  • Cas Number : 514787-47-4
  • Molecular Weight : 259.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018662.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018662.html
Availability details for
Quantity
  • Availability
  • Add to Cart