MAT089755851 Matrix Scientific

1-Cyclohexyl-1H-benzoimidazol-5-ylamine

  • MDL number MFCD02933874
  • Cas Number : 385381-36-2
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018670.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018670.html
Availability details for
Quantity
  • Availability
  • Add to Cart